L(+)-Arginine USP
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:74-79-3
:
Formula:C₆H₁₄N₄O₂
MDL Number:MFCD00002635
:174.2 g/mol
Merck Index:13,00785
Boiling Pt:368 °C (1013 hPa)
Melting Pt:238 °C
Density:1.3 g/cm³ (20 °C)
EINECS:200-811-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay:98.5% Min.
Specification Test Results
Frequently Bought Together