Ethyl 4-hydroxybenzoate NF
Supplier: Spectrum Chemical
About this item
Synonyms: Nipagin A, Ethyl paraben, Ethyl p-hydroxybenzoate, Ethylparaben, Ethyl 4-hydroxybenzoate
- CAS Number:120-47-8
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00002353
- Molecular Weight:166.18 g/mol
- Merck Index:13,03869
- Boiling Pt:297 °C (1013 hPa)
- Melting Pt:116…118 °C
- Density:1.078 g/cm³ (131 °C)
- Storage Temperature:Ambient
- EINECS:204-399-4
3 Options Available Below
Product Details & Documents
Synonyms: Nipagin A, Ethyl paraben, Ethyl p-hydroxybenzoate, Ethylparaben, Ethyl 4-hydroxybenzoate
- CAS Number:120-47-8
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00002353
- Molecular Weight:166.18 g/mol
- Merck Index:13,03869
- Boiling Pt:297 °C (1013 hPa)
- Melting Pt:116…118 °C
- Density:1.078 g/cm³ (131 °C)
- Storage Temperature:Ambient
- EINECS:204-399-4
Specifications for Product Family
Specification Test Results
- Assay:98% Min.
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



