Order Entry
Switzerland
Orders LinkContactUsLinkComponent
2-Ethoxyethyl acetate | Avantor

2-Ethoxyethyl acetate

Entzündlich
Gesundheit
Reizend
Anbieter:   Spectrum Chemical
2-Ethoxyethyl acetate
Anbieter:  Spectrum Chemical
Zertifikate

3 Varianten

Product Details & Documents
Synonyme:  Ethylglycol acetate
  • CAS Nummer:111-15-9
  • Formel:C₆H₁₂O₃
  • MDL-Nummer:MFCD00009251
  • Molekulargewicht:132.16 g/mol
  • Merck-Index:13,03787
  • Siedepunkt:156 °C (1013 hPa)
  • Schmelzender Platingehalt:–62 °C
  • Dichte:0.976 g/cm³ (20 °C)
  • Blitz Pt:54 °C
  • EINECS:203-839-2
  • UN:1172
  • ADR:3,III
Spezifikationen für diese Produktfamilie

Ergebnisse des Spezifikationstests

  • Assay:
    99% Min.
Product Family Options

Produktinformationen

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.