DL-Valine ethyl ester hydrochloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:23358-42-1
:
Formula:C₇H₁₅NO₂·HCl
MDL Number:MFCD00067542
:181.7 g/mol
Melting Pt:∼ 94 °C
Storage Temperature:Ambient
EINECS:245-606-8
Related Information
Frequently Bought Together