Order Entry
Switzerland
ContactUsLinkComponent
Celite AW Standard Supercel®, filter aid AW
Health
Irritant
Celite AW Standard Supercel®, filter aid AW
Catalog # L14552.36
CAS Number:  
Health
Irritant
Celite AW Standard Supercel®, filter aid AW
Catalog # L14552.36
CAS Number:  

Specifications

  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

CAS Number:61790-53-2
 :
MDL Number:MFCD00132803
Merck Index:14,4973
EINECS:319-127-6
 :GHS08
 :GHS07
Storage Temperature:Ambient