Order Entry
Switzerland
ContactUsLinkComponent
 
Phosphate-buffered saline (DPBS, 1X), Dulbecco's formula, without calcium, without magnesium
 
Phosphate-buffered saline (DPBS, 1X), Dulbecco's formula, without calcium, without magnesium
 

 

  • MDL No.:
    MFCD00131855
  • Physical form:
    Liquid
  • Appearance:
    Colourless

 

Frequently Bought Together