Order Entry
Switzerland
ContactUsLinkComponent
HEPES lysis buffer with NP-40 (2X)
HEPES lysis buffer with NP-40 (2X)
Catalog # J63867.AK
CAS Number:  
HEPES lysis buffer with NP-40 (2X)
Catalog # J63867.AK
CAS Number:  

Specifications

  • Pk:
    250 mL
  • Description:
    HEPES lysis buffer with NP-40 (2X)
  • Physical form:
    Liquid
  • Appearance:
    Clear liquid

Specifications

About this item

Physical Form: Liquid
Appearance: Clear liquid