Succinate, 0,2 M buffer solution, pH 5,0
Supplier: Thermo Fisher Scientific
About this item
2 Options Available Below
Product Details & Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- J63863.AK
- J63863.AP
- Odour/scent:
- Odourless
- Odourless
- CAS No.:
- 150-90-3
- 150-90-3
- MDL No.:
- MFCD00002789
- MFCD00002789
- Appearance:
- Colourless
- Colourless
- Physical form:
- Liquid
- Liquid
- Storage temperature:
- 2°C to 8°C
- 2°C to 8°C
Specifications
Product Family Options
Product Information
- PkAvailabilityPrice
- J63863.AKSuccinate, 0.2M buffer soln., pH 5.0Specifications:
More
Specifications
Cat. No.J63863.AKOdour/scentOdourlessCAS No.150-90-3MDL No.MFCD00002789AppearanceColourlessPhysical formLiquidStorage temperature2°C to 8°C - J63863.APSuccinate, 0.2M buffer soln., pH 5.0Specifications:
More
Specifications
Cat. No.J63863.APOdour/scentOdourlessCAS No.150-90-3MDL No.MFCD00002789AppearanceColourlessPhysical formLiquidStorage temperature2°C to 8°C



