Order Entry
Switzerland
ContactUsLinkComponent
TRIS-buffered saline (TBS, 20X), with 0,5% Tween 20
TRIS-buffered saline (TBS, 20X), with 0,5% Tween 20
Certificates
TRIS-buffered saline (TBS, 20X), with 0,5% Tween 20
Certificates

Specifications

  • MDL No.:
    MFCD00678198
  • Physical form:
    Liquid

Specifications

Related Information

Frequently Bought Together