

Sulindac Sulphide
Catalog # J62104.MC
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
CAS Number:32004-67-4
:
Formula:C₂₀H₁₇FO₂S
MDL Number:MFCD00869764
:340.4 g/mol
Melting Pt:189…191 °C
EINECS:250-892-2
:GHS08
:GHS07
Storage Temperature:Ambient