Phosphate, 0,5M buffer solution, pH 8,0
Supplier: Thermo Fisher Scientific
About this item
CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527
2 Options Available Below
Product Details & Documents
CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- J61722.AK
- J61722.AP
- Physical form:
- Liquid
- Liquid
- CAS No.:
- 7558-80-7
- 7558-80-7
- MDL No.:
- MFCD00003527
- MFCD00003527
Specifications
Product Family Options
Product Information
- ConcentrationpH valuePkAvailabilityPrice
- J61722.AKPhosphate buffer solnSpecifications:
More
Specifications
Cat. No.J61722.AKPhysical formLiquidCAS No.7558-80-7MDL No.MFCD00003527 - J61722.APPhosphate buffer solnSpecifications:
More
Specifications
Cat. No.J61722.APPhysical formLiquidCAS No.7558-80-7MDL No.MFCD00003527
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



