Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Thioflavin T ≥75%, tech.

Thioflavin T ≥75%, tech.

Thioflavin T ≥75%, tech.
Certificates
Thioflavin T ≥75%, tech.
Thioflavin T ≥75%, tech.

2 Options Available Below

Product Details & Documents

Purity: 75% CAS No.: 2390-54-7 Molecular Formula: C17H19ClN2S Formula Weight: 318.86 Physical Form: Powder Appearance: Yellow Beilstein Reference: 3922452 MDL No.: MFCD00011944

Applications: A dye used to visualize beta-amyloid plaques, like those associated with Alzheimer's Disease

Synonyms:  CI 49005, Thioflavine T, Basic Yellow 1, 2-(4-(Dimethylamino)phenyl)-3,6-dimethylbenzo[d]thiazol-3-ium chloride, Setoflavine T
  • CAS Number:2390-54-7
  • Formula:C₁₇H₁₉ClN₂S
  • MDL Number:MFCD00011944
  • Molecular Weight:318.86 g/mol
  • Storage Temperature:Ambient
  • EINECS:219-228-9
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.