Order Entry
Switzerland
Orders LinkContactUsLinkComponent
AMPD, 0.2 M Buffer Solutions

AMPD, 0.2 M Buffer Solutions

AMPD, 0.2 M Buffer Solutions
Certificates
AMPD, 0.2 M Buffer Solutions

2 Options Available Below

Product Details & Documents

2-Amino-2-methyl-1,3-propanediol buffer, colorless and odorless.

CAS No.: 115-69-5
MDL No.: MFCD00004678

Caution: Storage Temperature: 2 to 8 °C

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • J60916.AE
  • J60916.AK
  • Odour/scent:
  • Odourless
  • Odourless
  • Physical form:
  • Liquid
  • Liquid
  • CAS No.:
  • 115-69-5
  • 115-69-5
  • MDL No.:
  • MFCD00004678
  • MFCD00004678
  • Appearance:
  • Colourless
  • Colourless
  • Storage temperature:
  • 2°C to 8°C
  • 2°C to 8°C

Specifications

Product Family Options

Product Information

  • Concentration
    pH value
    Pk
    Availability
    Price
  • AMPD buffer solution
    0,2 M
    9,5
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - SFr. 103.00
    SFr. 103.00
    1 * 100 mL
       SFr. 103.00

    Specifications:
    More
    Specifications
    Cat. No.
    J60916.AE
    Odour/scent
    Odourless
    Physical form
    Liquid
    CAS No.
    115-69-5
    MDL No.
    MFCD00004678
    Appearance
    Colourless
    Storage temperature
    2°C to 8°C
  • AMPD buffer solution
    0,2 M
    9,5
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - SFr. 138.00
    SFr. 138.00
    1 * 250 mL
       SFr. 138.00

    Specifications:
    More
    Specifications
    Cat. No.
    J60916.AK
    Odour/scent
    Odourless
    Physical form
    Liquid
    CAS No.
    115-69-5
    MDL No.
    MFCD00004678
    Appearance
    Colourless
    Storage temperature
    2°C to 8°C