NP-40 lysis buffer
Supplier: Thermo Fisher Scientific
About this item
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
2 Options Available Below
Product Details & Documents
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- J60766.AP
- J60766.AK
- Appearance:
- Colourless
- Colourless
- Physical form:
- Liquid
- Liquid
- Storage temperature:
- 2°C to 8°C
- 2°C to 8°C
Specifications
Product Family Options
Product Information
- PkAvailabilityPrice
- J60766.APNP-40 lysis bufferSpecifications:
More
Specifications
Cat. No.J60766.APAppearanceColourlessPhysical formLiquidStorage temperature2°C to 8°C - J60766.AKNP-40 lysis bufferSpecifications:
More
Specifications
Cat. No.J60766.AKAppearanceColourlessPhysical formLiquidStorage temperature2°C to 8°C
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



