Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Phosphate, 0,2 M buffer solution, pH 7,0

Phosphate, 0,2 M buffer solution, pH 7,0

Phosphate, 0,2 M buffer solution, pH 7,0
Certificates
Phosphate, 0,2 M buffer solution, pH 7,0

2 Options Available Below

Product Details & Documents

CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • J60669.AP
  • J60669.K2
  • Physical form:
  • Liquid
  • Liquid
  • CAS No.:
  • 7558-80-7
  • 7558-80-7
  • MDL No.:
  • MFCD00003527
  • MFCD00003527

Specifications

Product Family Options

Product Information

  • Concentration
    pH value
    Pk
    Availability
    Price
  • J60669.AP
    Phosphate buffer soln
    Phosphate buffer soln
    0,2 M
    7,0
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - SFr. 125.00
    SFr. 125.00
    1 * 500 mL
       SFr. 125.00

    Specifications:
    More
    Specifications
    Cat. No.
    J60669.AP
    Physical form
    Liquid
    CAS No.
    7558-80-7
    MDL No.
    MFCD00003527
  • J60669.K2
    Phosphate buffer soln
    Phosphate buffer soln
    0,2 M
    7,0
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - SFr. 184.00
    SFr. 184.00
    1 * 1 L
       SFr. 184.00

    Specifications:
    More
    Specifications
    Cat. No.
    J60669.K2
    Physical form
    Liquid
    CAS No.
    7558-80-7
    MDL No.
    MFCD00003527