Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Phosphate, 0,2 M buffer solution, pH 8,0

Phosphate, 0,2 M buffer solution, pH 8,0

Phosphate buffer soln, 0,2 M, 8,0, 500 mL
Catalog # J60480.AP
Phosphate, 0,2 M buffer solution, pH 8,0
Catalog # J60480.AP
CAS Number:  
Phosphate, 0,2 M buffer solution, pH 8,0
Phosphate, 0,2 M buffer solution, pH 8,0

Phosphate buffer soln, 0,2 M, 8,0, 500 mL

CAS Number:  
Price
SFr. 131.00
Log in for availability
1 * 500 mL
Product Details & Documents

CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527

Specifications

Specifications

  • Concentration:
    0,2 M
  • Pk:
    500 mL
  • Description:
    Phosphate buffer soln
  • pH value:
    8,0
  • Cat. No.:
    J60480.AP
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00003527
  • CAS No.:
    7558-80-7

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.