Order Entry
Switzerland
ContactUsLinkComponent
NP-40 lysis buffer with glycerol (2X)
NP-40 lysis buffer with glycerol (2X)
Catalog # J60143.AK
CAS Number:  
NP-40 lysis buffer with glycerol (2X)
Catalog # J60143.AK
CAS Number:  

Specifications

  • Pk:
    250 mL
  • Description:
    NP-40 lysis buffer with glycerol (2X)
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Viscous liquid
  • Appearance:
    Colourless

Specifications

About this item

Storage Temperature: 2 to 8°C
Physical Form: Viscous liquid
Appearance: Colourless