Fig tree Ficin (from Latex)
Catalog # J60136.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 g
- CAS No.:9001-33-6
- Source:Latex
- Species:Fig tree
- Storage conditions:–30...–10 °C
- Form:Pale brown Lyophillized powder
- MDL No.:MFCD00081597
- Merck reference:14,4077
- Enzyme name:Ficin
Specifications
About this item
Applications: Proteolytic enzyme from fig latex, that is a cysteine endopeptidase