Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Denaturing lysis buffer

Denaturing lysis buffer

Denaturing lysis buffer, 250 mL
Catalog # J60132.AK
Denaturing lysis buffer
Catalog # J60132.AK
CAS Number:  
Denaturing lysis buffer
Denaturing lysis buffer

Denaturing lysis buffer, 250 mL

CAS Number:  
Price
SFr. 71.40
Log in for availability
1 * 250 mL
Product Details & Documents

Physical Form: Liquid
Appearance: Clear liquid
pH: 7,4

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    Denaturing lysis buffer
  • Cat. No.:
    J60132.AK
  • pH:
    7.4
  • Physical form:
    Liquid
  • Appearance:
    Clear liquid

Specifications