BIS-TRIS, 0,2 M Buffer Solutions
Supplier: Thermo Fisher Scientific
About this item
CAS No.: 6976-37-0
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
Odour: Odourless
MDL No.: MFCD00002853
8 Options Available Below
Product Details & Documents
CAS No.: 6976-37-0
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
Odour: Odourless
MDL No.: MFCD00002853
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- J62686.AE
- J62686.AK
- J60127.AE
- J60127.AK
- J63346.AE
- J63346.AK
- J62209.AE
- J62209.AK
- Odour/scent:
- Odourless
- Odourless
- Odourless
- Odourless
- Odourless
- Odourless
- Odourless
- Odourless
- Physical form:
- Liquid
- Liquid
- Liquid
- Liquid
- Liquid
- Liquid
- Liquid
- Liquid
- CAS No.:
- 6976-37-0
- 6976-37-0
- 6976-37-0
- 6976-37-0
- 6976-37-0
- 6976-37-0
- 6976-37-0
- 6976-37-0
- MDL No.:
- MFCD00002853
- MFCD00002853
- MFCD00002853
- MFCD00002853
- MFCD00002853
- MFCD00002853
- MFCD00002853
- MFCD00002853
- Appearance:
- Colourless
- Colourless
- Colourless
- Colourless
- Colourless
- Colourless
- Colourless
- Colourless
- Storage temperature:
- 2°C to 8°C
- 2°C to 8°C
- 2°C to 8°C
- 2°C to 8°C
- 2°C to 8°C
- 2°C to 8°C
- 2°C to 8°C
- 2°C to 8°C
Specifications
Product Family Options
Product Information
- ConcentrationpH valuePack typePkAvailabilityPrice
- J62686.AEBIS-TRIS buffer solutionSpecifications:
More
Specifications
Cat. No.J62686.AEOdour/scentOdourlessPhysical formLiquidCAS No.6976-37-0MDL No.MFCD00002853AppearanceColourlessStorage temperature2°C to 8°C - J62686.AKBIS-TRIS buffer solutionSpecifications:
More
Specifications
Cat. No.J62686.AKOdour/scentOdourlessPhysical formLiquidCAS No.6976-37-0MDL No.MFCD00002853AppearanceColourlessStorage temperature2°C to 8°C - J60127.AEBIS-TRIS buffer solutionSpecifications:
More
Specifications
Cat. No.J60127.AEOdour/scentOdourlessPhysical formLiquidCAS No.6976-37-0MDL No.MFCD00002853AppearanceColourlessStorage temperature2°C to 8°C - J60127.AKBIS-TRIS buffer solutionSpecifications:
More
Specifications
Cat. No.J60127.AKOdour/scentOdourlessPhysical formLiquidCAS No.6976-37-0MDL No.MFCD00002853AppearanceColourlessStorage temperature2°C to 8°C - J63346.AEBIS-TRIS buffer solutionSpecifications:
More
Specifications
Cat. No.J63346.AEOdour/scentOdourlessPhysical formLiquidCAS No.6976-37-0MDL No.MFCD00002853AppearanceColourlessStorage temperature2°C to 8°C - J63346.AKBIS-TRIS buffer solutionSpecifications:
More
Specifications
Cat. No.J63346.AKOdour/scentOdourlessPhysical formLiquidCAS No.6976-37-0MDL No.MFCD00002853AppearanceColourlessStorage temperature2°C to 8°C - J62209.AEBIS-TRIS buffer solutionSpecifications:
More
Specifications
Cat. No.J62209.AEOdour/scentOdourlessPhysical formLiquidCAS No.6976-37-0MDL No.MFCD00002853AppearanceColourlessStorage temperature2°C to 8°C - J62209.AKBIS-TRIS buffer solutionSpecifications:
More
Specifications
Cat. No.J62209.AKOdour/scentOdourlessPhysical formLiquidCAS No.6976-37-0MDL No.MFCD00002853AppearanceColourlessStorage temperature2°C to 8°C
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..



