Elastase substrate V
Catalog # J60093.MB
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 mg
- CAS No.:72252-90-5
- Storage conditions:–30...–10 °C
- Form:White powder
- MDL No.:MFCD00077135
- Enzyme name:Elastase Substrate V
- Solubility:Soluble in DMSO
- Molecular weight:627.70
- Molecular formula:C₃₁H₄₁N₅O₉
Specifications
About this item
Applications: Fluorogenic substrate for quantification of human leukocyte and porcine pancreatic elastase.