BES, 0.5 M buffer solutions
BES buffer solution, 0,5 M, 6,0, 250 mL
Nome del fornitore: Thermo Fisher Scientific
BES, 0.5 M buffer solutions
Codice catalogo J60058.AK
Nome del fornitore: Thermo Fisher Scientific
Numero CAS:
Vai a
Product Details & Documents
CAS No.: 66992-27-6
Storage temperature: 2 to 8 °C
Physical form: Liquid
Appearance: Colourless
MDL No.: MFCD00067425
Documents
Specifiche
Specifiche
- Concentration:0,5 M
- Pk:250 mL
- Description:BES buffer solution
- pH value:6,0
- Cat. No.:J60058.AK
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- MDL No.:MFCD00067425
- Appearance:Colourless
- CAS No.:66992-27-6



