DL-tert-Leucine (H-DL-Tle-OH) 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:33105-81-6
:
Formula:C₆H₁₃NO₂
MDL Number:MFCD00065933
:131.2 g/mol
Melting Pt:>300 °C (dec.)
Storage Temperature:Ambient
Related Information
Frequently Bought Together