
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt) ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:92921-24-9
:
Formula:C₁₆H₁₈N₂O₉S
MDL Number:MFCD00054978
:436.37 g/mol
:GHS07
Related Information
Frequently Bought Together