1-Phenyl-1-hexyn-3-ol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1817-51-2
:
Formula:C₁₂H₁₄O
MDL Number:MFCD00046687
:174.24 g/mol
Density:0.949
Flash Pt:110 °C (230 °F)
Storage Temperature:Ambient
Frequently Bought Together