Order Entry
Switzerland
ContactUsLinkComponent
Methyl-2-(trifluoromethyl)pyrimidine-4-carboxylate 97%
Irritant
Methyl-2-(trifluoromethyl)pyrimidine-4-carboxylate 97%
Catalog # H52818.03
CAS Number:  
Irritant
Methyl-2-(trifluoromethyl)pyrimidine-4-carboxylate 97%
Catalog # H52818.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:878745-51-8
 :
Formula:C₇H₅F₃N₂O₂
MDL Number:MFCD05662690
 :206.12 g/mol
 :GHS07
Storage Temperature:Ambient