Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt) 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:106627-54-7
:
Formula:C₄H₄NNaO₆S
MDL Number:MFCD00043100
:217.13 g/mol
Melting Pt:250 °C
Storage Temperature:Ambient
Related Information
Frequently Bought Together