Order Entry
Switzerland
ContactUsLinkComponent
 
 
Agarose 6% B - CL, cross-linked, beads
  H32568.AU
 :  
 
Agarose 6% B - CL, cross-linked, beads
  H32568.AU
 :  

 

  • Pk:
    1.000 mL
  • Seq:
    2865

 

 

 :
 :100 °C
 :1.040
 :38 °C (100 °F)
 :Refrigerator
 :1170
 :3,III
 :GHS02
 :9012-36-6
 :MFCD00081294