Order Entry
Switzerland
ContactUsLinkComponent
Agarose 4% B - CL, cross-linked, beads
Flammable
Agarose 4% B - CL, cross-linked, beads
Catalog # H32305.AU
CAS Number:  
Flammable
Agarose 4% B - CL, cross-linked, beads
Catalog # H32305.AU
CAS Number:  

Specifications

  • Pk:
    1.000 mL
  • Seq:
    2865

Specifications

About this item

 :
Boiling Pt:100 °C
Density:1.040
Flash Pt:38 °C (100 °F)
Storage Temperature:Refrigerator
UN:1170
ADR:3,III
 :GHS02
CAS Number:9012-36-6
MDL Number:MFCD00081294