2-Bromo-1-(pyridin-4-yl)ethanone hydrobromide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 4-(Bromoacetyl)pyridine hydrobromide
CAS Number:5349-17-7
:
Formula:C₇H₇Br₂NO
MDL Number:MFCD02681893
Melting Pt:∼ 200 °C (dec.)
ADR:8,II
:280.95 g/mol
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together