
BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester) 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:55750-62-4
:
Formula:C₁₁H₁₀N₂O₆
MDL Number:MFCD00043141
:266.21 g/mol
Melting Pt:168…170 °C
:GHS07
Related Information
Frequently Bought Together