

Boron trifluoridedimethanol complex (50 - 52% BF₃) (w/w)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2802-68-8
:373-57-9
Formula:BF₃·2CH₃OH
MDL Number:MFCD00071635
:131.89 g/mol
Boiling Pt:58…60 °C (4 mmHg)
Density:1.222
Flash Pt:68 °C (154 °F)
Storage Temperature:Ambient
EINECS:220-543-9
UN:2922
ADR:8,II
:GHS06
:GHS05
Related Information
Frequently Bought Together