
Sodium-2-naphthalenesulphonate 98% may cont. up to 10% residual inorganic salts and water
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:532-02-5
:
Formula:C₁₀H₈O₃S
MDL Number:MFCD00064186
:230.21 g/mol
EINECS:208-523-8
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together