Semiquantitative Standard 1, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Semiquantitative Standard 1, Specpure® is used as a standard for inductively coupled plasma and direct current plasma (DCP) emission spectrometry for the analysis of trace elements in biomedical and environmental samples.
UN No.: UN3093
MDL No.: MFCD02091742
Specifications
- MDL No.:MFCD02091742
Specifications
Frequently Bought Together