Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Aluminium oxide ≥99%

Aluminium oxide ≥99%
Certificates
Aluminium oxide ≥99%
Aluminium oxide ≥99%

2 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 32-63 Micron APS Powder, S.A. 200m2/g Density: 3.965 Refractive Index: 1.768 Appearance: White Odour: Odourless Melting Point: 2045° Boiling Point: 2980° MDL No.: MFCD00003424

Synonyms:  Aluminium(III) oxide, Alumina C, Aluminum oxide
  • CAS Number:1344-28-1
  • Formula:Al₂O₃
  • MDL Number:MFCD00003424
  • Molecular Weight:101.96 g/mol
  • Merck Index:14,356
  • Boiling Pt:2980 °C
  • Melting Pt:2045 °C
  • Density:3.965
  • Storage Temperature:Ambient
  • EINECS:215-691-6
Product Family Options

Product Information