


n-Hexane, anhydrous, over molecular sieves
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520
:86.18 g/mol
Merck Index:14,4694
Boiling Pt:69 °C
Melting Pt:–95 °C
Density:0.659
EINECS:203-777-6
UN:1208
ADR:3,II
:Packaged under argon in resealable ChemSeal™ bottles.
Flash Pt:–23 °C (–9 °F)
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
Frequently Bought Together