
Silicon ≥99.999% (metals basis), powder -325 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-21-3
:
Formula:Si
MDL Number:MFCD00085311
:28.09 g/mol
Storage Temperature:Ambient
EINECS:231-130-8
UN:1346
ADR:4.1,III
:GHS02
Related Information
Frequently Bought Together