Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Aluminium oxide basic for TLC

Aluminium oxide basic for TLC

Aluminium oxide basic for TLC
Certificates
Aluminium oxide basic for TLC
Aluminium oxide basic for TLC

2 Options Available Below

Product Details & Documents

CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 5-15 Micron APS Powder Refractive Index: 1.765 Appearance: White to pale pink Odour: Odourless Melting Point: 2045° Boiling Point: 2980° MDL No.: MFCD00003424

  • CAS Number:1344-28-1
  • Formula:Al₂O₃
  • MDL Number:MFCD00003424
  • Molecular Weight:101.96 g/mol
  • Merck Index:14,356
  • Boiling Pt:2980 °C
  • Melting Pt:2045 °C
  • Storage Temperature:Ambient
  • EINECS:215-691-6
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.