Order Entry
Switzerland
ContactUsLinkComponent
Zinc 68, 68 Zn, plasma standard solution, 68 Zn 10 µg/ml, Specpure®
Zinc 68, 68 Zn, plasma standard solution, 68 Zn 10 µg/ml, Specpure®
Catalog # 45289.AD
CAS Number:  
Zinc 68, 68 Zn, plasma standard solution, 68 Zn 10 µg/ml, Specpure®
Catalog # 45289.AD
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    50 mL
  • MDL No.:
    MFCD00145542
  • Element:
    Zinc
  • Symbol:
    Zn

Specifications

About this item

Molecular Formula: Matrix: 2% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00145542