Order Entry
Switzerland
ContactUsLinkComponent
 
Boron 11, 11 B, plasma standard solution, 11 B 100 µg/ml, Specpure®
  45273.AD
 :  
Boron 11, 11 B, plasma standard solution, 11 B 100 µg/ml, Specpure®
  45273.AD
 :  

 

  • Concentration:
    100 mg/L
  • Matrix:
    H₂O
  • Pk:
    50 mL
  • MDL No.:
    MFCD00151272
  • Element:
    Boron
  • Symbol:
    B

 

 

Molecular Formula: Matrix: H₂O
Physical Form: Liquid
MDL No.: MFCD00151272