Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), flakes -150 mesh, Thickness: 1.2 µm Type 316-L
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
Formula:Fe
MDL Number:MFCD00148499
:55.845 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together