Order Entry
Switzerland
ContactUsLinkComponent
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), spheres, Ø 3 mm (0.12 in) Type 316
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), spheres, Ø 3 mm (0.12 in) Type 316
Catalog # 42020.KM
CAS Number:  
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), spheres, Ø 3 mm (0.12 in) Type 316
Catalog # 42020.KM
CAS Number:  

Specifications

  • Pk:
    25
  • Seq:
    0025

Specifications

About this item

 :
 :55.845 g/mol
Storage Temperature:Ambient
Formula:Fe
MDL Number:MFCD00148499