Order Entry
Switzerland
ContactUsLinkComponent
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), spheres, Ø 2 mm (0.08 in) Type 316
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), spheres, Ø 2 mm (0.08 in) Type 316
Catalog # 42019.KN
CAS Number:  
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), spheres, Ø 2 mm (0.08 in) Type 316
Catalog # 42019.KN
CAS Number:  

Specifications

  • Pk:
    100
  • Seq:
    0100

Specifications

About this item

 :
 :55.845 g/mol
Storage Temperature:Ambient
Formula:Fe
MDL Number:MFCD00148499