Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 20 mesh woven from 0.382mm (0.015in) Type 304
Anbieter: Thermo Fisher Scientific
Zertifikate
Über diesen Artikel
:
Formel:Fe
MDL-Nummer:MFCD00148499
:55.845 g/mol
Lagertemperatur:Ambient
Frequently Bought Together