Sulphur in heavy mineral oil standard solutions, Specpure®
Specpure® calibration standards, designed specifically for the analysis of sulphur in petroleum products by X-ray fluorescence spectrometry.
Physical Form: Liquid
MDL No.: MFCD00804270
- MDL No.:MFCD00804270
- Physical form:Liquid
Frequently Bought Together