Order Entry
Switzerland
ContactUsLinkComponent
Multi-element QC-7 check standard solution, Specpure®
Multi-element QC-7 check standard solution, Specpure®
Catalog # 39145.AE
CAS Number:  
Multi-element QC-7 check standard solution, Specpure®
Catalog # 39145.AE
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃/tr HF
  • Pk:
    100 mL
  • Description:
    Multi-Element QC-7 Check Standard Solution
  • MDL No.:
    MFCD00210654

Specifications

About this item

UN No.: UN3264
MDL No.: MFCD00210654