Orders LinkContactUsLinkComponent
Ceramic plates, rectangular or square, AL23
Ceramic plates, rectangular or square, AL23
Ceramic plates, rectangular or square, AL23
Certificates
Ceramic plates, rectangular or square, AL23
3 Options Available Below
Product Details & Documents
Pure Al₂O₃ Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.
Product Family Options
Product Information
  • Thickness
    Width
    Length
    Availability
    Price
  • 4 mm
    25 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 127.00
    SFr. 127.00
    1 * 1 ST
       SFr. 127.00
  • 4 mm
    50 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 201.00
    SFr. 201.00
    1 * 1 ST
       SFr. 201.00
  • 4 mm
    50 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 358.00
    SFr. 358.00
    1 * 1 ST
       SFr. 358.00