Order Entry
Switzerland
ContactUsLinkComponent
Stainless steel (Fe:Cr:Ni; 73:18:9 wt%), rods, Ø 6.35 mm (0.25 in) Type 303
Stainless steel (Fe:Cr:Ni; 73:18:9 wt%), rods, Ø 6.35 mm (0.25 in) Type 303
Catalog # 13473.BV
CAS Number:  
Stainless steel (Fe:Cr:Ni; 73:18:9 wt%), rods, Ø 6.35 mm (0.25 in) Type 303
Catalog # 13473.BV
CAS Number:  

Specifications

  • Pk:
    30 cm
  • Seq:
    7765

Specifications

About this item

 :
 :55.845 g/mol
Storage Temperature:Ambient
Formula:Fe
MDL Number:MFCD00148499