Order Entry
Switzerland
ContactUsLinkComponent
pH/mV/°C/ISE meter, bench, Lab 845
pH/mV/°C/ISE meter, bench, Lab 845
Catalog # 663-0323
Supplier:  SI Analytics
undefined
pH/mV/°C/ISE meter, bench, Lab 845
Catalog # 663-0323
Supplier:  SI Analytics

Specifications

  • Temperature range:
    −10...+100 °C
  • Description:
    pH/mV/°C/ISE meter, bench, Lab 845 (meter only without electrode)
  • Seq:
    01
  • Model:
    Lab 845
  • Temperature accuracy:
    ± 0,1 (0...100 °C)
  • Temperature resolution:
    0,1 °C
  • Connections:
    BNC, 2 × banana socket (4 mm)
  • Data logging capacity:
    4000 entries with date, time, value 1+2 and temperature
  • Display:
    Graphic LCD, 128×64 pixel, with backlight
  • ISE range:
    0 - 30000 ppm
  • Outputs:
    USB, isolated
  • Temperature compensation:
    Automatic with Pt1000 or fix temperature
  • Weight:
    0,75 kg (incl. power supply and stand)
  • W×D×H:
    145×55×185 mm
  • mV accuracy:
    ± 0,3
  • mV range:
    ±1999
  • mV resolution:
    1
  • pH accuracy:
    ± 0,01 (± 2 pH around calibration point)
  • pH calibration:
    2-point (Automatic)
  • pH range:
    0 - 14
  • pH resolution:
    0,01
  • Pk:
    1

Specifications

About this item

This small benchtop meter with intuitive operation and robust aluminum housing is perfect for training and routine purposes.

  • User-friendly design, numerical access protection possible
  • Graphic backlight LCD display
  • Data logger (4000 data sets)
  • Min/Max values with date and time stamp
  • Aluminum desk housing (IP 40/DIN EN 60529)
  • Developed and made in Germany
Delivery: Meter only is supplied with power supply.
Lab 845 Set/BL19pH also includes stand, pH electrode BlueLine 19 pH, and DIN buffers in ampules (6 pieces).
Lab 845 Set/BL25pH also includes stand, pH electrode BlueLine 25 pH, and DIN buffers in ampules (6 pieces).
Lab 845 Set/BL29pH also includes stand, pH electrode BlueLine 29 pH, and DIN buffers in ampules (6 pieces).