ContactUsLinkComponent
Test kits, arsenic, Spectroquant®, Supelco®
Test kits, arsenic, Spectroquant®, Supelco®
Catalog # 1.01747.0001
Supplier:  Merck
CAS Number:  
undefined
Test kits, arsenic, Spectroquant®, Supelco®
Catalog # 1.01747.0001
Supplier:  Merck
CAS Number:  
Specifications
  • Chemical method:
    Silver DDTC
  • Pk:
    30 Tests
  • Description:
    Arsenic test*²
  • Seq:
    14
  • M-Clarity quality level:
    MQ100
  • For unit:
    Prove, Nova 60, Multy, Move
  • Range:
    0,001 - 0,100 mg/l As
Specifications
About this item
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Highly stable, ready to use reagent mixtures in a user friendly design
  • Free selection of appropriate cell format according to the measuring range
  • Long shelf life up to 3 years - all at room temperature
  • Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*2 Arsenic reagents 2 and 7, 1.00731.1000 and 1.08780.0500 together with 1.73501.0001 required